missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SNRPD1 Polyclonal specifically detects SNRPD1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | SNRPD1 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | HsT2456, small nuclear ribonucleoprotein D1 polypeptide 16kDa, small nuclear ribonucleoprotein Sm D1, Sm-D autoantigen, Sm-D1, SMD1, snRNP core protein D1, SNRPD |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-119 of human SNRPD1 (NP_008869.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?