missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10352-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects 13-16 kDa (SMD1) and 30 kDa (SMD1/SMD2 dimer form).
SNRPD1 is a small nuclear ribonucleoprotein that belongs to the SNRNP core protein family. This protein may act as a charged protein scaffold to promote SNRNP assembly or strengthen SNRNP-SNRNP interactions through nonspecific electrostatic contacts with RNA.Specifications
| SNRPD1 | |
| Polyclonal | |
| Unconjugated | |
| SNRPD1 | |
| HsT2456, Sm D autoantigen, Sm D1, SMD1, snRNP core protein D1, SNRPD, SNRPD1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20641, 291794, 6632 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62314, P62315 | |
| Snrpd1 | |
| SNRPD1 Fusion Protein Ag0364 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction