missing translation for 'onlineSavingsMsg'
Learn More

SNRPD1 Antibody, Biotin, Novus Biologicals™

Product Code. 18373065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373065 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373065 Supplier Novus Biologicals Supplier No. NBP306017B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNRPD1 Polyclonal specifically detects SNRPD1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPD1
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot
Formulation PBS
Gene Alias HsT2456, small nuclear ribonucleoprotein D1 polypeptide 16kDa, small nuclear ribonucleoprotein Sm D1, Sm-D autoantigen, Sm-D1, SMD1, snRNP core protein D1, SNRPD
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SNRPD1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6632
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.