missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRPC Recombinant Rabbit Monoclonal Antibody (K01_2J74)

Product Code. 30828806
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828806 50 μL
30829374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828806 Supplier Invitrogen™ Supplier No. MA565365
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

SNRPC Recombinant Monoclonal Antibody for Western Blot

SNRPC is associated with snRNP U1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNRPC
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2J74
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene SNRPC
Gene Accession No. D3ZCL3, P09234
Gene Alias small nuclear ribonucleoprotein polypeptide C; Snrp1c; Snrpc; U1 small nuclear ribonucleoprotein 1C; U1 small nuclear ribonucleoprotein C; U1 small nuclear RNP specific C; U1 snRNP C; U1 snRNP protein C; U1C; U1-C; U1C protein; Yhc1
Gene Symbols SNRPC
Host Species Rabbit
Immunogen A synthetic peptide of human U1-C.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361808, 6631
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.