missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRPB2 Monoclonal Antibody (OTI2E8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526941
This item is not returnable.
View return policy
Description
SNRPB2 Monoclonal Antibody for Western Blot, ICC/IF
SNRPB2 is involved in pre-mRNA splicing. This protein is associated with snRNP U2. It binds stem loop IV of U2 snRNA only in presence of the U2A' protein.
Specifications
| SNRPB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08579 | |
| SNRPB2 | |
| Full length human recombinant protein of SNRPB2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| OTI2E8 | |
| Unconjugated | |
| SNRPB2 | |
| 2810052G09Rik; Msl1; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptide B''; small nuclear ribonucleoprotein polypeptide B2; Snrpb2; U2 small nuclear ribonucleoprotein B; U2 small nuclear ribonucleoprotein B''; U2 snRNP B''; U2B'' | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6629 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction