missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNRPB Monoclonal Antibody (Y12)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA513449

Product Code. 11314853

  • £500.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SNRPB Monoclonal Antibody for ICC/IF, IHC (P), IM, RIP, Western Blot

The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B') have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNRPB
Monoclonal
200 μg/mL
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
P14678, P27048
SNRPB
Protein G
RUO
100341600, 100624586, 171365, 20638, 6628
4°C
Liquid
Immunohistochemistry (Paraffin), Immunomicroscopy, Western Blot, RNA Immunoprecipitation, Immunocytochemistry
Y12
Unconjugated
SNRPB
AL024368; AU018828; B polypeptide of Sm protein; CCMS; COD; sm protein B; Sm protein B/B'; SM11; small nuclear ribonucleoprotein B; small nuclear ribonucleoprotein polypeptide B; small nuclear ribonucleoprotein polypeptides B and B'; small nuclear ribonucleoprotein polypeptides B and B1; small nuclear ribonucleoprotein-associated protein B; Small nuclear ribonucleoprotein-associated proteins B and B'; smB; sm-B; Sm-B/B'; SmB/B'; sm-B/Sm-B'; SmB/SmB'; SNRNP-B; Snrpb; SNRPB1
Mouse
500 μL
Primary
Drosophila, Human, Mouse, Pig, Rabbit, Rat
Antibody
IgG3 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.