missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35914-20ul
This item is not returnable.
View return policy
Description
SNRPB Polyclonal antibody specifically detects SNRPB in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SNRPB | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CODB polypeptide of Sm protein, Sm protein B/B', small nuclear ribonucleoprotein polypeptides B and B', small nuclear ribonucleoprotein polypeptides B and B1, small nuclear ribonucleoprotein-associated proteins B and B', SmB/B', Sm-B/B'sm-B/Sm-B', SmB/SmB', snRNP-Bsmall nuclear ribonucleoprotein polypeptide B, SNRPB1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human SNRPB (NP_003082.1).,, Sequence:, MTVGKSSKMLQHIDYRMRCILQDGRIFIGTFKAFDKHMNLILCDCDEFRKIKPKNSKQAEREEKRVLGLVLLRGENLVSMTVEGPPPKDTGIARVPLAGA | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6628 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur