missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRP70 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-57487
This item is not returnable.
View return policy
Description
SNRP70 Polyclonal specifically detects SNRP70 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SNRP70 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| RNPU1ZU170K, RPU1U1AP, small nuclear ribonucleoprotein 70kDa (U1), Snp1, snRNP70, SNRP70U1 small nuclear ribonucleoprotein 70 kDa, U1 snRNP 70 kDa, U1-70Ksmall nuclear ribonucleoprotein 70kDa (RNP antigen), U1AP1, U1RNP | |
| Rabbit | |
| 51 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Human: 100%; Western clawed frog: 100%; Bovine: 100%; Zebrafish: 100%; Xenopus: 100%; Canine: 100%; Rat: 100%; African malaria mosquito: 92%; Yellowfever mosquito: 92%; Mosquito: 92%; Harpegnathos saltator: 92%; Camponotus floridanus: 92%; Brown alga: 92%; Red flour beetle: 92%; Florida lancelet: 92%; Black-legged tick: 92%; Trichoplax reptans: 92%; Fruit fly: 92%; Caenorhabditis elegans: 85%; Caenorhabditis vulgaris: 85%; Eye worm: 85%; Body louse: 85%; Caenorhabditis briggsae: 85%; Filarial nematode worm: 85%; Planarian: 85%; Starlet sea anemone: 85%; Toxoplasma gondii ME49: 84%; Toxoplasma gondii: 84%; Blood fluke: 83%; Moss: 76%; Spikemoss: 76%; Volvox carteri f. nagariensis: 76%; Chlorella variabilis: 76%; Chlamydomonas smithii: 76%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml | |
| P08621 | |
| SNRNP70 | |
| Synthetic peptides corresponding to SNRP70 (small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen)) The peptide sequence was selected from the N terminal of SNRP70 (NP_003080). Peptide sequence PHNDPNAQGDAFKTLFVARVNYDTTESKLRREFEVYGPIKRIHMVYSKRS The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 6625 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction