missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRNP70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110406
This item is not returnable.
View return policy
Description
Immunogen sequence: EYEHERDMHS AYKHADGKKI DGRRVLVDVE RGRTVKGWRP RRLGGGLGGT RRGGADVNIR HSGRDDTSRY DERPGPSPLP HRDRDRDRER ERRERSRERD K.
SNRP70 contains 1 RRM (RNA recognition motif) domain and mediates the splicing of pre-mRNA by binding to the loop I region of U1-snRNA. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications
| SNRNP70 | |
| Polyclonal | |
| Unconjugated | |
| SNRNP70 | |
| 2700022N21Rik; 3200002N22Rik; AI325098; LOW QUALITY PROTEIN: U1 small nuclear ribonucleoprotein 70 kDa; R74807; RNPU1Z; Rnu1p70; Rnulp70; RPU1; small nuclear ribonucleoprotein 70 (U1); small nuclear ribonucleoprotein 70kDa; small nuclear ribonucleoprotein 70kDa (U1); small nuclear ribonucleoprotein 70kDa polypeptide (RNP antigen); small nuclear ribonucleoprotein U1 subunit 70; small nuclear ribonucleoprotein, U1 70kDa subunit; Snp1; Snrnp70; SNRP70; snrp70 protein; Srnp70; U1 small nuclear ribonucleoprotein 70 kDa; U1 small nuclear ribonucleoprotein polypeptide A; U1 small ribonucleoprotein 70kd; U1 snRNP 70 kDa; U1-70; U170K; U1-70K; U1AP; U1AP1; U1RNP; Unknown (protein for MGC:138058); wu:fc20b04; zgc:136450 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20637, 361574, 6625 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08621, Q62376 | |
| SNRNP70 | |
| A synthetic peptide corresponding to a sequence within amino acids 150-250 of human SNRNP70 (NP_003080.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction