missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRNP35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564304
This item is not returnable.
View return policy
Description
Immunogen sequence: LGGGLGGKKE SGQLRFGGRD RPFRKPINLP VVKNDLYREG KRERRERS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
U1SNRNPBP is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors.The protein encoded by this gene is a homolog of U1-snRNP binding protein. Its N-terminal half contains a RNA recognition motif and the C-terminal half is rich in Arg/Asp and Arg/Glu dipeptides, a characteristic of a variety of splicing factors. This gene is differentially expressed in a variety of human tissues. Multiple alternatively spliced transcript variants have been found for this gene, and they differ in the 5' sequence regions.
Specifications
| SNRNP35 | |
| Polyclonal | |
| Unconjugated | |
| SNRNP35 | |
| 6330548G22Rik; HM1; HM-1; Protein HM-1; RGD1310724; small nuclear ribonucleoprotein 35 (U11/U12); small nuclear ribonucleoprotein 35kDa (U11/U12); small nuclear ribonucleoprotein U11/U12 subunit 35; small nuclear ribonucleoprotein, U11/U12 35kDa subunit; SNRNP35; U1 snRNP binding protein homolog; U1 snRNP-binding protein homolog; U11/U12 small nuclear ribonucleoprotein 35 kDa protein; U11/U12 snRNP 35 kDa protein; U11/U12 snRNP 35K; U11/U12-35K; U1-snRNP binding protein homolog; U1snrnpbp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11066 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16560 | |
| SNRNP35 | |
| Recombinant protein fragment of Human SNRNP35 (Product #RP-103647) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction