missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRNP27 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83568-0.1ML
This item is not returnable.
View return policy
Description
SNRNP27 Polyclonal specifically detects SNRNP27 in Mouse samples. It is validated for Western Blot.
Specifications
| SNRNP27 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 11017 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Nucleic acid-binding protein RY-1, putative nucleic acid binding protein RY-1, RY1,27K, small nuclear ribonucleoprotein 27kDa (U4/U6.U5), U4/U6.U5 small nuclear ribonucleoprotein 27 kDa protein, U4/U6.U5 snRNP 27 kDa protein, U4/U6.U5 tri-snRNP-associated 27 kDa protein, U4/U6.U5 tri-snRNP-associated protein 3, U4/U6.U5-27K | |
| The immunogen is a synthetic peptide directed towards the middle region of mouse SNRNP27 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction