missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRNP25 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560658
This item is not returnable.
View return policy
Description
Immunogen sequence: EEALPHSEAM DVFQEGLAMV VQDPLLCDLP IQVTLEEVNS QIALEYGQAM TVRVCKMDGE V Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.
Specifications
| SNRNP25 | |
| Polyclonal | |
| Unconjugated | |
| SNRNP25 | |
| C16orf33; minus-99 protein; small nuclear ribonucleoprotein 25kDa (U11/U12); small nuclear ribonucleoprotein U11/U12 subunit 25; small nuclear ribonucleoprotein, U11/U12 25kDa subunit; SNRNP25; U11/U12 small nuclear ribonucleoprotein 25 kDa protein; U11/U12 snRNP 25 kDa protein; U11/U12 snRNP 25K | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79622 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BV90 | |
| SNRNP25 | |
| Recombinant protein corresponding to Human SNRNP25. Recombinant protein control fragment (Product #RP-98709). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction