missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRNP25 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93543-0.1ml
This item is not returnable.
View return policy
Description
SNRNP25 Polyclonal antibody specifically detects SNRNP25 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SNRNP25 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| C16orf33, chromosome 16 open reading frame 33, FLJ22940, Minus-99 protein, small nuclear ribonucleoprotein 25kDa (U11/U12), U11/U12 small nuclear ribonucleoprotein 25 kDa protein, U11/U12 snRNP 25 kDa protein, U11/U12 snRNP 25K, U11/U12-25K | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-132 of human SNRNP25 (NP_078847.1). MDVFQEGLAMVVQDPLLCDLPIQVTLEEVNSQIALEYGQAMTVRVCKMDGEVMPVVVVQSATVLDLKKAIQRYVQLKQEREGGIQHISWSYVWRTYHLTSAGEKLTEDRKKLRDYGIRNRDEVSFIKKLRQK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79622 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction