Learn More
SNRK Polyclonal Antibody
GREENER_CHOICE
SNRK Polyclonal Antibody
Brand:  Invitrogen PA544539
Product Code. 15546532
  • £444.00 / 100µL
Description
SNRK Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNRK
Polyclonal
Unconjugated
SNRK
2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312
Rabbit
Affinity Chromatography
RUO
54861
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9NRH2
SNRK
Synthetic peptide directed towards the middle region of human SNRK.