missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SnoN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106469
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SKIL.
TGF-beta is a ubiquitously expressed cytokine that signals through the Smad protein family to regulate numerous cellular processes such as embryonic development and tumorigenesis. This signaling is negatively regulated by Ski, the mammalian homolog of the transforming protein of an avian retrovirus that induces oncogenic transformation of chicken embryo cells, and the related protein SnoN. Like Ski, SnoN functions by binding to the Smad proteins and preventing their phosphorylation, thereby inhibiting their ability to bind DNA and activate the transcription of downstream genes. SnoN is located primarily in the nucleus in cancer tissues or cells, but in the cytoplasm in normal tissues or primary epithelial cells.
Specifications
| SnoN | |
| Polyclonal | |
| Unconjugated | |
| SKIL | |
| EGK_11909; LOW QUALITY PROTEIN: ski-like protein; SKI like proto-oncogene; ski/sno related; SKIL; SKI-like; SKI-like oncogene; ski-like protein; ski-like protein-like; SKI-like proto-oncogene; Skir; ski-related oncogene; ski-related oncogene snoN; Ski-related protein; SNO; SnoA; SnoI; SnoN; v-ski sarcoma viral oncogene homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114208, 20482, 6498 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12757, Q60665 | |
| SKIL | |
| A synthesized peptide derived from human SKIL(Accession P12757), corresponding to amino acid residues Q632-A682. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction