missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SnoN Antibody (OTI3E2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74255AF405
This item is not returnable.
View return policy
Description
SnoN Monoclonal specifically detects SnoN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SnoN | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SKIL | |
| Recombinant protein fragment corresponding to amino acids 307-684 of human SnoN (NP_005405) produced in E.coli. | |
| 0.1 mL | |
| 6498.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI3E2 | |
| Western Blot, Immunohistochemistry | |
| SKI-like oncogene, Ski-related oncogene, ski-related oncogene snoN, Ski-related protein, SnoA, SnoI, SnoN, SNOski-like protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction