missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SnoN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38200-20ul
This item is not returnable.
View return policy
Description
SnoN Polyclonal antibody specifically detects SnoN in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SnoN | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| SKI-like oncogene, Ski-related oncogene, ski-related oncogene snoN, Ski-related protein, SnoA, SnoI, SnoN, SNOski-like protein | |
| A synthetic peptide corresponding to a sequence within amino acids 350-450 of human SnoN (NP_005405.2).,, Sequence:, EMKEKFSMRSGKRNQSKTDAPSGMELQSWYPVIKQEGDHVSQTHSFLHPSYYLYMCDKVVAPNVSLTSAVSQSKELTKTEASKSISRQSEKAHSSGKLQKT | |
| 20 μL | |
| Cancer, Signal Transduction | |
| 6498 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction