missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14950-1-AP-20UL
This item is not returnable.
View return policy
Description
Smad-binding peptide aptamers can be developed to selectively inhibit TGF-beta-induced gene expression.Specifications
| SNIP1 | |
| Polyclonal | |
| Unconjugated | |
| SNIP1 | |
| RP3 423B22.3, SNIP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 313588, 76793, 79753 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M9G6, Q8BIZ6, Q8TAD8 | |
| Snip1 | |
| SNIP1 Fusion Protein Ag6772 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction