missing translation for 'onlineSavingsMsg'
Learn More

SNF5 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-57525CL1

Product Code. 30155223

  • £771.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SNF5 Polyclonal antibody specifically detects SNF5 in Human,Mouse samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNF5
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
CoraFluor 1
BAF47, BAF47hSNF5, BRG1-associated factor 47, hSNFS, Ini1, INI1RTPS1, Integrase interactor 1 protein, malignant rhabdoid tumor suppressor, RDTSNF5 homolog, Sfh1p, SNF5, SNF5L1, Snr1, subfamily b, member 1, sucrose nonfermenting, yeast, homolog-like 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily B member 1
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human SMARCB1/SNF5 using the numbering given in entry NP_003064.2 (GeneID 6598).
0.1 mL
Cell Cycle and Replication
6598
Store at 4°C in the dark. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.