missing translation for 'onlineSavingsMsg'
Learn More

SNF1LK2/SIK2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18408023 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408023 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408023 Supplier Novus Biologicals Supplier No. NBP142690DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SNF1LK2/SIK2 Polyclonal antibody specifically detects SNF1LK2/SIK2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNF1LK2/SIK2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SNF1-like kinase 2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 600 and 650 of human salt-inducible kinase 2 using the numbering given in entry NP_056006.1 (GeneID 23235).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 23235
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.