missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SNED1 Polyclonal specifically detects SNED1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | SNED1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | DKFZp586B2420, FLJ00133,6720455I24Rik homolog, insulin responsive sequence DNA binding protein-1, Insulin-responsive sequence DNA-binding protein 1, IRE-BP1, Snep, SST3, sushi, nidogen and EGF-like domain-containing protein 1, sushi, nidogen and EGF-like domains 1 |
| Gene Symbols | SNED1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:GVRVSIRHPEALRDQATDVDRSVDRFTFRALLPGKRYTIQLTTLSGLRGEEHPTESLATAPTHVWTRPLPPANLTAARVTATSA |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?