Learn More
SND1 Mouse anti-Human, CoraLite 594, Clone: 1A6A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60265-100UL
Product Code. 17204796
  • £375.00 / 100µL
Description
Staphylococcal nuclease domain-containing 1 (SND1), is a multifunctional nuclease that consists of four staphylococcal nuclease domains and a tudor domain. SND1 acts as a coactivator that facilitates transcriptional activity of STAT5, 6 and c-Myc. SND1 is a comprising part of the RNA-induced silencing complex(RISC), and takes part in the functions of miRNA, regulates transcription through transcriptional coactivation, RNA interference, RNA splicing, and RNA editing. Higher level of SND1 has been found in colon cancer and prostate cancer, can promote HCC angiogenesis in xenograft model through induction of angiogenic factors.
Specifications
Specifications
SND1
Monoclonal
1000 μg/mL
Mouse
Primary
Human
IgG1
Immunofluorescence
1A6A4
CoraLite 594
100 μL
27044