missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ SNaPshot™ Multiplex Kit

Product Code. 15829766 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 Reactions
100 Reactions, with protocol
1000 Reactions
5000 Reactions
Unit Size:
100 reactions
1000 reactions
5000 reactions
Each
This item is not returnable. View return policy

Product Code. 15829766

Brand: Applied Biosystems™ 4323163

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SNaPshot™ Multiplex Ready Reaction Mix (for 5000 reactions), SNaPshot™ Multiplex Control Primer Mix (30μL), SNaPshot™ Multiplex Control Template (60μL)

The SNaPshot™ Multiplex Kit is part of the SNaPshot™ Multiplex System. The SNaPshot™ Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs).

The SNaPshot® Multiplex Kit is part of the SNaPshot® Multiplex System. The SNaPshot® Multiplex System primer extension-based method enables multiplexing up to 10 single nucleotide polymorphisms (SNPs). Use this system to screen and confirm SNPs, detect minor sequence variations, assess DNA methylation, fingerprint bacterial artificial chromosomes (BACs) or general labeling of restriction fragments.

• Interrogate up to 10 SNPs from different amplicons in a single base extension reaction
• Separate SNP loci that differ by a single base pair
• Increased sensitivity compared to standard sequencing allows detection of minor variants (e.g. somatic mutations) in a test sample
• Reduce the complexity of your experiments with this easy to use and cost-efficient genotyping system
• A large body of scientifc literature demonstrates the utility for genotyping, mutation detection and mapping

The SNaPshot® Multiplex Kit is a part of the SNaPshot® Multiplex System:
1) SNaPshot® Multiplex Kit (100, 1000, 5000 reaction sizes)
2) GeneScan™-120 LIZ™ Size Standard
3) Matrix Standard Set DS-02 for dye set E5 appropriate for your instrument

Each of the three components above is required to run the SNaPshot® Multiplex Kit. The Matrix Standard set, however, is only required at a one-time initial set up to calibrate the instrument for the SNaPshot-specific dye set E5.

4) GeneMapper® Software v.4.0 (or higher version) is the recommended secondary data analysis software that enables set up of user-defined panels and bins for automated data analysis.

Alternatively, simple, manual peak analysis is possible using PeakScanner™ 1.0 software (free download from website) for the 310, 3100. 3130, and 3730 instruments (PeakScanner is not compatible with data from 3500 instruments). For the 3500 instruments the primary data analysis software on the instrument allows fragment sizing and color separation similar to PeakScanner.

5) The SNaPshot primer Focus Kit (4329538) is a useful accessory kit for single experiment or assay (panel) design: Analyze individual primers for their approximate sizing location. This will determine whether or not two oligonucleotides will produce overlapping signals when run simultaneously and enable the setting of proper loci windows (bins) for GeneMapper analysis software.

Screen and Confirm SNPs
The SNaPshot® Multiplex System is the perfect tool for SNP screening and validation. The kit offers a one-tube single-base extension⁄termination reagent to label DNA fragments. Run DNA fragments on any Applied Biosystems capillary electrophoresis instrument with the GeneScan™-120 LIZ® Size Standard to indicate the size of labeled fragments. Then use GeneMapper® Software to analyze the data and generate allele calls. The SNaPshot® Multiplex Kit allows multiplexing during single-base extension of up to 10 primer-template combinations in a single tube-single capillary format.

Assess DNA Methylation
The study of methylation⁄epigenetics is emerging as an important component of cancer research. In a typical assay to detect methylation, bisulfite treatment of DNA deaminates non-methylated cytosine and converts it to uracil while methylated cytosine remains unreactive. The subsequent step of PCR amplification coverts uracil bases to thymine. Use SNaPshot® todetect the base differences in treated and untreated samples to learn the methylation status of your samples.

Fingerprint BACs
New BAC libraries require a rapid, efficient method for characterization (also called fingerprinting) and assembly of clones into contigs (contiguous consensus sequences), which are then arrayed into physical maps of the chromosome. Use the SNaPshot® Multiplex Kit to fingerprint clones by labeling BAC fragments after digestion with restriction endonucleases. The labeled fragments can then be separated and detected on any Applied Biosystems capillary electrophoresis instrument. Sizing information from GeneMapper® Software v 4.0 or higher is imported into subsequent editing and contig assembly programs.

In a similar fashion, any restriction fragment with a 5' overhang can be labeled with a fluorescent dye using the SNaPshot reagent.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains:
• SNaPshot™ Multiplex Ready Reaction Mix (for 5000 reactions)
• SNaPshot™ Multiplex Control Primer Mix (30 μl)
• SNaPshot™ Multiplex Control Template (60 μl)

Store at -15°C to -25°C in a constant-temperature freezer.
Detection Method Primer-probe
Format Tube(s)
Polymerase AmpliTaq DNA Polymerase
Technique SNP Genotyping (Fragment Analysis-Based), Primer Extension
For Use With (Equipment) 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer, 3730/3730xl Genetic Analyzers
Genotyping Target SNPs (Known)
High-throughput Compatibility Multiplexing
Product Line SNaPshot
Product Type Multiplex Kit
Quantity 5,000 reactions
Shipping Condition Dry Ice
Software GeneMapper™ Software
Hot Start No
No. of Reactions 5000 Reactions
Sample Type DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.