missing translation for 'onlineSavingsMsg'
Learn More

SNAPIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen SNAPIN
Concentration 0.35 mg/mL
Applications Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16846203
View Documents
Proteintech
10055-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16836203
View Documents
Proteintech
10055-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody can recognize the 15-18 kDa (Monomer) and 30-36 kDa (Dimer) forms of SNAPIN.

The protein encoded by this gene is a coiled-coil-forming protein that associates with the SNARE (soluble N-ethylmaleimide-sensitive fusion protein attachment protein receptor) complex of proteins and the BLOC-1 (biogenesis of lysosome-related organelles) complex. Biochemical studies have identified additional binding partners. As part of the SNARE complex, it is required for vesicle docking and fusion and regulates neurotransmitter release. The BLOC-1 complex is required for the biogenesis of specialized organelles such as melanosomes and platelet dense granules. Mutations in gene products that form the BLOC-1 complex have been identified in mouse strains that are models of Hermansky-Pudlak syndrome. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

SNAPIN
Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
Unconjugated
Rabbit
Rat, Mouse, Human
O95295, P60192, Q9Z266
20615, 23557, 295217
SNAPIN Fusion Protein Ag0101
Primary
-20°C
SNAPIN
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
SNAP associated protein, SNAP25BP, SNAPAP, SNAPIN
Snapin
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.