missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAPC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538551
Code : NEW
Additional Details : Weight : 0.00900kg
Specifications
SNAPC5 | |
Polyclonal | |
Unconjugated | |
SNAPC5 | |
SNAP19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10302, 330959 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75971, Q8R2K7 | |
SNAPC5 | |
A synthetic peptide derived from the N-terminal region of human SNAPC5 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |