missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAPC5 Monoclonal Antibody (5C3)

Description
Peptide Sequence: MLSRLQELRK EEETLLRLKA ALHDQLNRLK MLVHVDNEAS INQTTLELST KSHVTEEEEE EEEEESDS.
Part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. Binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. Recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
Specifications
| Antigen | SNAPC5 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 5C3 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SNAPC5 |
| Gene Accession No. | O75971 |
| Gene Alias | 2010103A03Rik; CTD-3185P2.1; small nuclear RNA activating complex polypeptide 5; small nuclear RNA activating complex, polypeptide 5; small nuclear RNA activating complex, polypeptide 5, 19kD; small nuclear RNA activating complex, polypeptide 5, 19kDa; small nuclear RNA-activating complex polypeptide 5; SNAP19; SNAPc 19 kDa subunit; SNAPc subunit 5; SNAPC5; snRNA-activating protein complex 19 kDa subunit; snRNA-activating protein complex subunit 5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?