missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAPC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567278
This item is not returnable.
View return policy
Description
Immunogen sequence: MDVDAEREKIT QEIKELERIL DPGSSGSHVE ISESSLESDS EADSLPSEDL DPADPPISEE ERWGEASNDE DDPKDKTLPE DPETCLQLNM VYQEVIQE Highest antigen sequence identity to the following orthologs - mouse 78%, rat 76%.
Part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. Binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. Recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
| SNAPC4 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC4 | |
| GTAR; KIAA0697; proximal sequence element-binding transcription factor subunit alpha; PSE-binding factor subunit alpha; PTF subunit alpha; PTFalpha; small nuclear RNA activating complex polypeptide 4; small nuclear RNA activating complex, polypeptide 4; small nuclear RNA activating complex, polypeptide 4, 190kD; small nuclear RNA activating complex, polypeptide 4, 190kDa; SNAP190; SNAPc 190 kDa subunit; SNAPc subunit 4; SNAPC4; snRNA-activating protein complex 190 kDa subunit; snRNA-activating protein complex subunit 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SXM2 | |
| SNAPC4 | |
| Recombinant Human SNAPC4. Recombinant protein control fragment (Product #RP-107915). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction