missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SNAPC4 Monoclonal antibody specifically detects SNAPC4 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
Specifications
Specifications
| Antigen | SNAPC4 |
| Applications | ELISA, Western Blot, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 1G2 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_003077 |
| Gene Alias | FLJ13451, Proximal sequence element-binding transcription factor subunit alpha, PSE-binding factor subunit alpha, PTF subunit alpha, PTFalpha, small nuclear RNA activating complex, polypeptide 4, 190kD, small nuclear RNA activating complex, polypeptide 4, 190kDa, SNAP190SNAPc 190 kDa subunit, SNAPc subunit 4, snRNA-activating protein complex 190 kDa subunit, snRNA-activating protein complex subunit 4 |
| Host Species | Mouse |
| Immunogen | SNAPC4 (NP_003077, 53 a.a. ∽ 162 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PADPPISEEERWGEASNDEDDPKDKTLPEDPETCLQLNMVYQEVIQEKLAEANLLLAQNREQQEELMRDLAGSKGTKVKDGKSLPPSTYMGHFMKPYFKDKVTGVGPPAN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?