missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAPC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27062-1-AP-150UL
This item is not returnable.
View return policy
Description
SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.Specifications
| SNAPC3 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC3 | |
| PTF subunit beta, PTFbeta, SNAP50, SNAPc 50 kDa subunit, SNAPc subunit 3, SNAPC3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6619 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92966 | |
| SNAPC3 | |
| SNAPC3 Fusion Protein Ag19851 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction