missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAPC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100779
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SNAPC3.
SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
| SNAPC3 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC3 | |
| 1810020H02Rik; 4930558A07Rik; 5031401C21Rik; AI414457; E030018J20Rik; proximal sequence element-binding transcription factor subunit beta; PSE-binding factor subunit beta; PTF subunit beta; PTFbeta; small nuclear RNA activating complex polypeptide 3; small nuclear RNA activating complex, polypeptide 3; small nuclear RNA activating complex, polypeptide 3, 50kD; small nuclear RNA activating complex, polypeptide 3, 50kDa; small nuclear RNA-activating complex polypeptide 3; SNAP50; SNAPc 50 kDa subunit; SNAPc subunit 3; SNAPC3; snRNA-activating protein complex 50 kDa subunit; snRNA-activating protein complex subunit 3; Unknown (protein for MGC:133802) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6619 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92966 | |
| SNAPC3 | |
| A synthesized peptide derived from human SNAPC3(Accession Q92966), corresponding to amino acid residues N368-N411. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction