missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAPC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571460
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
This gene encodes a subunit of the snRNA-activating protein complex which is associated with the TATA box-binding protein. The encoded protein is necessary for RNA polymerase II and III dependent small-nuclear RNA gene transcription. Alternative splicing results in multiple transcript variants.
Specifications
| SNAPC2 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC2 | |
| 0610007H10Rik; AU015675; Proximal sequence element-binding transcription factor subunit delta; PSE-binding factor subunit delta; PTF subunit delta; PTFDELTA; small nuclear RNA activating complex polypeptide 2; small nuclear RNA activating complex, polypeptide 2; small nuclear RNA activating complex, polypeptide 2, 45kD; small nuclear RNA activating complex, polypeptide 2, 45kDa; small nuclear RNA-activating complex polypeptide 2; SNAP45; SNAPc 45 kDa subunit; SNAPc subunit 2; SNAPC2; snRNA-activating protein complex 45 kDa subunit; snRNA-activating protein complex subunit 2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6618 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1.0 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13487 | |
| SNAPC2 | |
| synthetic peptide directed towards the middle region of human SNAPC2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction