missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAPC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584892
This item is not returnable.
View return policy
Description
Immunogen sequence: TPPGLQTDCE ALLSRFQETD SVRFEDFTEL WRNMKFGTIF CGRMRNLEKN MFTKEALALA WR Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 84%.
SNAPC1 is a part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. It binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes and recruits TBP and BRF2 to the U6 snRNA TATA box.
Specifications
| SNAPC1 | |
| Polyclonal | |
| Unconjugated | |
| SNAPC1 | |
| 2700033G17Rik; AU015787; Proximal sequence element-binding transcription factor subunit gamma; PSE-binding factor subunit gamma; PTF subunit gamma; PTFgamma; small nuclear RNA activating complex polypeptide 1; small nuclear RNA activating complex, polypeptide 1; small nuclear RNA activating complex, polypeptide 1, 43kD; small nuclear RNA activating complex, polypeptide 1, 43kDa; small nuclear RNA-activating complex polypeptide 1; SNAP43; SNAPc 43 kDa subunit; SNAPc subunit 1; SNAPC1; snRNA-activating protein complex 43 kDa subunit; snRNA-activating protein complex subunit 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6617 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16533 | |
| SNAPC1 | |
| Recombinant protein corresponding to Human SNAPC1. Recombinant protein control fragment (Product #RP-108597). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction