missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAP47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555782
This item is not returnable.
View return policy
Description
Immunogen sequence: DDIKVHSPYE ISIRQRFIGK PDMAYRLISA KMPEVIPILE VQFSKKMELL EDALVLRSAR TSSPAEKSCS VWHAASGLMG RTLHREPPAG DQE Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 81%.
SNAP47 is a protein coding gene.
Specifications
| SNAP47 | |
| Polyclonal | |
| Unconjugated | |
| Snap47 | |
| 1110031B06Rik; C1orf142; C7H1orf142; CH1073-162B20.3; epididymis luminal protein 170; epididymis secretory protein Li 290; ESFI5812; HEL170; HEL-S-290; LOW QUALITY PROTEIN: synaptosomal-associated protein 47; MGC72593; RGD735194; snap47; SNAP-47; SVAP1; Synaptosomal-associated 47 kDa protein; synaptosomal-associated protein 47; synaptosomal-associated protein, 47; synaptosomal-associated protein, 47kDa; synaptosome associated protein 47; synaptosome associated protein 47kDa; Unknown (protein for MGC:153222); zgc:153222 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SQN1 | |
| Snap47 | |
| Recombinant protein corresponding to Human SNAP47. Recombinant protein control fragment (Product #RP-94298). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction