missing translation for 'onlineSavingsMsg'
Learn More

SNAP45 Antibody (4B8), Novus Biologicals™

Product Code. 18383639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383639 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383639 Supplier Novus Biologicals Supplier No. H00006618M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SNAP45 Monoclonal antibody specifically detects SNAP45 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP45
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003074
Gene Alias Proximal sequence element-binding transcription factor subunit delta, PSE-binding factor subunit delta, PTF subunit delta, PTFdelta, small nuclear RNA activating complex, polypeptide 2, 45kD, small nuclear RNA activating complex, polypeptide 2, 45kDa, Small nuclear RNA-activating complex polypeptide 2, SNAP45snRNA-activating protein complex 45 kDa subunit, SNAPc 45 kDa subunit, SNAPc subunit 2, snRNA-activating protein complex subunit 2
Host Species Mouse
Immunogen SNAPC2 (NP_003074, 16 a.a. ~ 114 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GEVTGPATWSAREKRQLVRLLQARQGQPEPDATELARELRGRSEAEIRVFLQQLKGRVAREAIQKVHPGGLQGPRRREAQPPAPIEVWTDLAEKITGPL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6618
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.