missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP43 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93475-0.02ml
This item is not returnable.
View return policy
Description
SNAP43 Polyclonal antibody specifically detects SNAP43 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SNAP43 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Proximal sequence element-binding transcription factor subunit gamma, PSE-binding factor subunit gamma, PTF subunit gamma, PTFgamma, small nuclear RNA activating complex, polypeptide 1, 43kD, small nuclear RNA activating complex, polypeptide 1, 43kDa, Small nuclear RNA-activating complex polypeptide 1, SNAP43SNAPc 43 kDa subunit, SNAPc subunit 1, snRNA-activating protein complex 43 kDa subunit, snRNA-activating protein complex subunit 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-368 of human SNAPC1 (NP_003073.1). MGTPPGLQTDCEALLSRFQETDSVRFEDFTELWRNMKFGTIFCGRMRNLEKNMFTKEALALAWRYFLPPYTFQIRVGALYLLYGLYNTQLCQPKQKIRVALKDWDEVLKFQQDLVNAQHFDAAYIFRKLRLDRAFHFTAMPKLLSYRMKKKIHRAEVTEEFKDPSDRVMKLITSDVLEEMLNVHDHYQNMKHVISVDKSKPDKALSLIKDDFFDNIKNIVLEHQQWHKDRKNPSLKSKTNDGEEKMEGNSQETERCERAESLAKIKSKAFSVVIQASKSRRHRQVKLDSSDSDSASGQGQVKATRKKEKKERLKPAGRKMSLRNKGNVQNIHKEDKPLSLSMPVITEEEENESLSGTEFTASKKRRKH | |
| 0.02 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6617 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction