missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12704-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene, a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.Specifications
| SNAP29 | |
| Polyclonal | |
| Unconjugated | |
| SNAP29 | |
| CEDNIK, SNAP 29, SNAP29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116500, 67474, 9342 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95721, Q9ERB0, Q9Z2P6 | |
| SNAP29 | |
| SNAP29 Fusion Protein Ag3382 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction