missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Antibody (SP12), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81057AF350
This item is not returnable.
View return policy
Description
SNAP25 Monoclonal specifically detects SNAP25 in Human, Mouse, Rat, Porcine, Bovine, Feline, Guinea Pig, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SNAP25 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| SNAP25 | |
| Recombinant monoclonal antibody to SNAP25. | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 6616 | |
| Human, Mouse, Rat, Pig, Bovine, Feline, Guinea Pig, Hamster | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| SP12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to native and denatured SNAP25. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction