missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-737
This item is not returnable.
View return policy
Description
SNAP25 Polyclonal specifically detects SNAP25 in Rat samples. It is validated for Western Blot.
Specifications
| SNAP25 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| P60881 | |
| SNAP25 | |
| Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # P60881 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for the ∼25k SNAP25 protein. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Neuroscience, Neurotransmission, Vision | |
| 6616 | |
| Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction