missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10825-1-AP-20UL
This item is not returnable.
View return policy
Description
Synaptosomal-associated proteins (SNAPs) are cytosolic proteins that play a key role in in the process of membrane fusion in intracellular vesicle trafficking. In eukaryotic cells, the SNARE (soluble N-ethylmaleimide-sensitive factor attachment protein receptor) complex is critical to membrane docking and fusion and is believed to impart some degree of specificity between vesicle SNARE (v-SNARE) and target organelle SNARE (t-SNARE). In neurons and neuroendocrine cells, the SNARE complex consists of the integral membrane proteins VAMP (vesicle-associated membrane protein), syntaxin and SNAP-25. In non-neuronal tissue, a SNAP-25 homolog, SNAP-23, functionally replaces SNAP-25 in the SNARE complex. Studies show that VAMP, syntaxin and SNAP-23 are required for SNARE function and that this complex exists as a heterotrimer of the three proteins. In insulin signaling pathways, studies suggest that the translocation of GLUT4 in adipocytes requires that functional SNAP-23 be present in SNARE complexes.Specifications
| SNAP23 | |
| Polyclonal | |
| Unconjugated | |
| SNAP23 | |
| HsT17016, SNAP 23, SNAP23, SNAP23A, SNAP23B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20619, 64630, 8773 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00161, O09044, O70377 | |
| Snap23 | |
| SNAP23 Fusion Protein Ag1269 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction