missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Snap Tubes

Product Code. 15879117
Click to view available options
Volume (Metric) Well:
15 mL
50 mL
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 15879117

Brand: Macherey-Nagel™ 740823.50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Conical centrifuge tubes with snap lids for use as lysis tubes for incubation of samples in the purification of nucleic acids with compatible kits (e.g. NucleoSnap Plasmid Midi or NucleoMag DNA Plasma). Packs of 50 tubes.

Specifications

Color Clear
Format Conical Tube
For Use With (Equipment) Centrifuge
For Use With (Application) Incubation and lysis of samples
Includes Snap lid
Product Type Centrifuge Tube
Quantity 50 Pack
Volume (Metric) Well 15 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.