missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNAP Tag Polyclonal Antibody

Product Code. 10586290
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10586290 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10586290 Supplier Invitrogen™ Supplier No. CAB4255

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CAB4255 detects SNAP in samples. CAB4255 has been successfully used in Western Blot applications. The CAB4255 immunogen is purified recombinant SNAP.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SNAP Tag
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation BBS with no preservative; pH 8.4
Host Species Rabbit
Immunogen purified recombinant SNAP.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.