missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Snap Caps with Single Thumb Tab For 16mm Tubes

Product Code. 31922563 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Blue
Clear
Gray
Green
Lavender
Red
White
Yellow
Unit Size:
Pack of 1000
Pack of 10000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31922563 Clear Pack of 10000
31922751 Gray Pack of 10000
31922746 Blue Pack of 10000
31923273 Green Pack of 1000
31923449 Lavender Pack of 1000
31922806 Red Pack of 1000
31922510 White Pack of 1000
31922553 Yellow Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 31922563 Supplier Globe Scientific Supplier No. 113148C
Upgrade Your Laboratory Essentials with Globe Scientific
Use promo code 27959
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

To recap blood drawing tubes, glass test tubes and plastic culture tubes

  • Made of Low-density polyethylene
  • Maintains tight seal during centrifugation, agitation and refrigeration procedures
  • One thumb tab allows more tubes to be stacked closely and for easy removal of cap
  • Assorted colors available for sample identification purposes

Specifications

Includes Single thumb tab
Product Type Snap Cap
Material LDPE
Color Clear
For Use With (Equipment) 16 mm glass and evacuated blood collection tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.