missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAP alpha Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583808
This item is not returnable.
View return policy
Description
Immunogen sequence: IEEACEIYAR AANMFKMAKN WSAAGNAFCQ AAQLHLQLQS KHD Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
Specifications
| SNAP alpha | |
| Polyclonal | |
| Unconjugated | |
| NAPA | |
| 1500039N14Rik; alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; a-SNAP; AW209189; hydrocephaly with hop gait; hyh; Napa; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; NSF attachment protein alpha; RA81; Snap; SNAPA; SNAP-alpha; SNARE | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P54920 | |
| NAPA | |
| Recombinant protein corresponding to Human NAPA. Recombinant protein control fragment (Product #RP-98823). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction