missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNAP alpha Monoclonal Antibody (15D4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA112453
This item is not returnable.
View return policy
Description
SNAP alpha Monoclonal Antibody for Western Blot, IP, ELISA
SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs (SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-to-endosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
Specifications
| SNAP alpha | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and no preservative; pH 7.5 | |
| P54920, P54921, P81125 | |
| NAPA | |
| Recombinant human alpha-SNAP. | |
| 100 μg | |
| Primary | |
| Human, Rat, Bovine | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| 15D4 | |
| Unconjugated | |
| NAPA | |
| 1500039N14Rik; alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; a-SNAP; AW209189; hydrocephaly with hop gait; hyh; Napa; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; NSF attachment protein alpha; RA81; Snap; SNAPA; SNAP-alpha; SNARE | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 140673, 518407, 8775 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction