missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNAP alpha/beta Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA580840

Product Code. 16307924

  • £289.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SNAP-alpha.

SNAPs (soluble NSF attachment proteins), acting in concert with SNAREs ( SNAP receptors) and the N-ethylmaleimide-sensitive fusion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-toendosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SNAP alpha/beta
Polyclonal
Unconjugated
NAPA
alpha snap; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; bA416N4.2; CMS18; dJ1068F16.2; Napa; N-ethylmaleimide sensitive fusion protein attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; NSF attachment protein alpha; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; SNAP; SNAP25; SNAP-25; SNAP-25a; SNAP-25B; Snapa; SNAP-alpha; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25kDa
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
8775
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with no preservative
P54920
NAPA
Recombinant Human SNAP-alpha Protein.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.