missing translation for 'onlineSavingsMsg'
Learn More

SNAP 94847 hydrochloride, Tocris Bioscience™

Product Code. 13462854
Encompass
Click to view available options
10mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective MCH1 antagonist

Potent melanin-concentrating hormone receptor 1 (MCH1) antagonist (Ki = 2.2 nM, KD = 530 pM) that displays > 80-fold and > 500-fold selectivity over α1A and D2 receptors respectively. Increases neurogenesis in the dentate gyrus and decreases food-reinforced operant responding in vivo. Exhibits anxiolytic activity and is orally active.

Specifications

CAS 487051-12-7
Molecular Formula C29H33ClF2N2O2
Quantity 10mg
Synonym snap 94847 hydrochloride, n-3-1-4-3,4-difluorophenoxy phenyl methyl-4-piperidinyl-4-methylphenyl-2-methylpropanamide hydrochloride
InChI Key DEDUDFNRQKUBRH-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)NC(=O)C(C)C)C2CCN(CC2)CC3=CC=C(C=C3)OC4=CC(=C(C=C4)F)F.Cl
IUPAC Name N-[3-[1-[[4-(3,4-difluorophenoxy)phenyl]methyl]piperidin-4-yl]-4-methylphenyl]-2-methylpropanamide;hydrochloride
Molecular Weight (g/mol) 515.042
PubChem CID 56972235
Formula Weight 515.03
Percent Purity >98%
Chemical Name or Material SNAP 94847 hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'