missing translation for 'onlineSavingsMsg'
Learn More

Snail Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18429135 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429135 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429135 Supplier Novus Biologicals Supplier No. NBP227293AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Snail Polyclonal antibody specifically detects Snail in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Snail
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias dJ710H13.1, Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAI1, SNAIL, snail 1 homolog, snail 1 zinc finger protein, snail family zinc finger 1, snail homolog 1, SNAIL1, zinc finger protein SNAI1
Host Species Rabbit
Immunogen A portion of amino acids 1-60 of human SNAI1 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6615
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.