Learn More
SNAI2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12129-1-AP-150UL
Description
The SNAI2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNAI2. This antibody recognizes human, mouse antigen. The SNAI2 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
| SNAI2 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 6591 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |