missing translation for 'onlineSavingsMsg'
Learn More

sn-Glycerol 3-phosphate, MedChemExpress

Product Code. 16471189
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16471189

Brand: Medchem Express HY113128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress sn-Glycerol 3-phosphate is produced by cytosolic glycerol 3-phosphate dehydrogenase pathway through the reduction of dihydroxyacetone phosphate using NADH formed during glycolysis.

sn-Glycerol 3-phosphate is produced by cytosolic glycerol 3-phosphate dehydrogenase pathway through the reduction of dihydroxyacetone phosphate using NADH formed during glycolysis.

Specifications

Chemical Name or Material sn-Glycerol 3-phosphate
Physical Form Solid
CAS 17989-41-2
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C3H9O6P
Solubility Information H2O : 125 mg/mL (726.45 mM; Need ultrasonic)
SMILES O[C@H](CO)COP(O)(O)=O
Molecular Weight (g/mol) 172.07
Formula Weight 172.07
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.