missing translation for 'onlineSavingsMsg'
Learn More

sn-Glycerol 3-Phosphate Bis(cyclohexylammonium) Salt, TRC

Product Code. 30006080
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006080 250 mg 250mg
30012833 10 mg 10mg
30005269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006080 Supplier Toronto Research Chemicals Supplier No. TRCG601590250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29849-82-9
Molecular Formula C15H35N2O6P
Molecular Weight (g/mol) 370.42
Synonym (R)-1,2,3-Propanetriol 1-(Dihydrogen phosphate) compd. with cyclohexanamine, D-Glycerol 1-(Dihydrogen phosphate) compd. with cyclohexylamine, sn-Glycerol-3-phosphate Dicyclohexylammonium Salt
IUPAC Name cyclohexanamine;[(2R)-2,3-dihydroxypropyl] dihydrogen phosphate
SMILES NC1CCCCC1.NC2CCCCC2.OC[C@@H](O)COP(=O)(O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/2C6H13N.C3H9O6P/c2*7-6-4-2-1-3-5-6;4-1-3(5)2-9-10(6,7)8/h2*6H,1-5,7H2;3-5H,1-2H2,(H2,6,7,8)/t;;3-/m..1/s1
Formula Weight 370.2233
Chemical Name or Material sn-Glycerol 3-Phosphate Bis(cyclohexylammonium) Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.